Learn More
Medchemexpress LLC DL-lysine monohydrochloride | 70-53-1 | MFCD00064563 | 97.0% | 182.65 g/mol | C6H15ClN2O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW027251100G
Encompass_Preferred
DL-lysine monohydrochloride is the hydrochloride salt of the amino acid lysine, supplied as a solid reagent for laboratory and biochemical research. It is commonly used in buffer preparation, cell culture supplementation, and biochemical assays where lysine or its salt form is required. The product is provided with purity specifications and supporting documentation such as a certificate of analysis and safety data sheet.
  • Hydrochloride salt of DL-lysine for improved solubility in aqueous media.
  • High purity (97.0%) suitable for analytical and research applications.
  • Solid form, easy to handle, weigh, and store.
  • Molecular weight 182.65 g/mol; formula C6H15ClN2O2.
  • Supplied with certificate of analysis and safety data sheet for quality assurance.
  • Suitable for buffer formulation, cell culture supplementation, and biochemical assays.
Catalog No. 50-003-00307