Learn More
TARGETMOL CHEMICALS INC DL-MAACKIAIN 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6S021420MG
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. dl-Maackiain (Demethylpterocarpin) induces apoptosis and growth suppression 2. MAK1 is the second functional gene cloned from Maackiain detoxification. purity: 97%
Catalog No. 50-257-6390