Learn More
Medchemexpress LLC DL-methyldopa-d3 | 99.1% | 214.23 g/mol | C10H10D3NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY121809S5MG
Encompass_Preferred
DL-methyldopa-d3 is a deuterium-labeled analogue of DL-methyldopa intended for use as a tracer and as an internal standard for quantitative analytical methods, including NMR, GC-MS, and LC-MS. It is supplied as a solid with high purity and storage stability suitable for research and analytical workflows.
  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity (99.14%) appropriate for analytical applications.
  • Solid, white to off-white appearance for easy handling and weighing.
  • Stable under recommended storage conditions for reliable long-term use.
  • Suitable as a tracer in pharmacokinetic and metabolic studies.
Catalog No. 50-002-64510