Learn More
Medchemexpress LLC DL-Norepinephrine-d6 hydrochloride | 1219803-04-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN7142S310MG
Encompass_Preferred
DL-Norepinephrine-d6 hydrochloride is a deuterium-labeled form of DL-Norepinephrine hydrochloride. DL-Norepinephrine hydrochloride is an external racemic catecholamine neurotransmitter, consisting of an equal mixture of left-handed (L-, active) and right-handed (D-, very low or inactive) stereoisomers. This compound can be used as a tracer for Parkinson's disease research when labeled with tritium. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is utilized for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • May influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-19876