Learn More
Medchemexpress LLC DL-Norepinephrine-d6 (hydrochloride) | 1219803-04-9 | MFCD08705108 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN7142S35MG
Encompass_Preferred
DL-Norepinephrine-d6 hydrochloride is a deuterium labeled external racemic catecholamine neurotransmitter, identified as a marker for catecholamine-secreting tumors. It functions as an internal standard for quantitative analysis and can be used as a tracer in research, including for Parkinson's disease.
  • Deuterium labeled compound
  • Functions as an external racemic catecholamine neurotransmitter
  • Suitable for use as a tracer in research
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Useful for monitoring norepinephrine levels in urine or plasma
  • Modulates human dendritic cell activation by altering cytokine release
  • Acts as an antagonist of dibutyryl cyclic AMP in narcosis regulation
Catalog No. 50-004-19754