Learn More
Medchemexpress LLC HY-W010510 5g Medchemexpress, DL-Norvaline CAS:760-78-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0105105G
Encompass_Preferred
Medchemexpress, HY-W010510 5g DL-Norvaline CAS:760-78-1 DL-Norvaline, a derivative of L-norvaline , L-norvaline is a non-competitive inhibitor of arginase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4539