Learn More
Cayman Chemical DL-o-TyrosIn 250mg
SDP
Supplier:  Cayman Chemical 33570250 MG
Encompass_Preferred
A phenylalanine hydroxylation product; has been used as a marker of irradiation processing in poultry meat
Catalog No. 50-294-7252