Learn More
Medchemexpress LLC Dl-o-tyrosine | 2370-61-8 | 98.2% | 181.19 g/mol | C9H11NO3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10784810MM 1ML
Encompass_Preferred
DL-O-Tyrosine is an ortho-tyrosine derivative used as an amino acid reagent for biochemical research and assay development. It is offered as a solid and as a ready-to-use 10 mM solution in DMSO for convenient dosing in experimental workflows.
  • Available as solid (100 mg-1 g) and 10 mM solution in DMSO.
  • High purity: 98.23%.
  • Molecular formula C9H11NO3; molecular weight 181.19 g/mol.
  • White to pink solid appearance.
  • Supplied with COA and SDS documentation.
Catalog No. 50-002-44498