Learn More
Apexbio Technology LLC DL-Panthenol 16485-10-2 500mg
SDP
Supplier:  Apexbio Technology LLC C6776500
Encompass_Preferred
DL-Panthenol (CAS 16485-10-2) is an alcohol derivative of pantothenic acid (vitamin B5) It is designed to support epithelial cell metabolism by serving as a prodrug that is converted to pantothenic acid and subsequently integrated into Coenzyme A (CoA) DL-Panthenol exerts its biological activity primarily through its conversion into pantothenic acid in epithelial cells enhancing CoA-dependent metabolic processes This promotes fibroblast proliferation and re-epithelialization during wound repair No specific IC50/EC50 values or experimental models are provided Based on these pharmacological properties DL-Panthenol holds research potential in studies of epithelial regeneration wound healing and inflammation modulation in dermatology
Catalog No. 50-000-89813