Learn More
Medchemexpress LLC DL-penicillamine | 52-66-4 | MFCD00004856 | 98.0% | 149.21 g/mol | C5H11NO2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0174575MG
Encompass_Preferred
DL-Penicillamine (±-Penicillamine) is a low-molecular-weight thiol compound and copper-chelating agent supplied as a solid with high purity for biochemical and pharmacological research. It is commonly used as a tool compound in studies of metal chelation, heavy-metal toxicity models, and disorders of copper metabolism.
  • High purity suitable for research applications.
  • Solid form for precise dosing and formulation.
  • Functions as a copper chelator in biochemical assays.
  • Used in pharmacology and toxicology studies and models.
  • Compatible with common aqueous solvents for dissolution.
Catalog No. 50-002-99977