Learn More
Medchemexpress LLC DL-penicillamine | 52-66-4 | MFCD00004856 | 98.0% | 149.21 g/mol | C5H11NO2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01745750MG
Encompass_Preferred
DL-penicillamine is a small-molecule copper chelator used in biochemical and pharmacological research. It selectively binds copper and other heavy metals and is used in detoxification studies, biochemical assays, and mechanistic investigations. Supplied as a high-purity solid suitable for analytical and preparative applications.
  • High purity suitable for analytical use.
  • Effective copper chelating activity in biochemical assays.
  • Useful in heavy metal detoxification and toxicity studies.
  • Available as a solid reagent in a range of pack sizes.
  • Soluble in water for solution preparation.
Catalog No. 50-003-00408