Learn More
Cayman Chemical DL-Phenyl-d5-alnIn 1g
SDP
Supplier:  Cayman Chemical 348451 G
Encompass_Preferred
An internal standard for the quantification of DL-phenylalanine by GC- or LC-MS
Catalog No. 50-291-1000