Learn More
Indofine Chemical Dl-Phenylalanine, 100 Gm
SDP
Supplier:  Indofine Chemical 025872 100 GM
Encompass_Preferred
DL-PHENYLALANINE 100 GM, Amino Acids & Peptides, DL-Phe-OH, C9H11NO2, MW: 165.20, >99% Assay, 150-30-1, MFCD00064225
Catalog No. 50-909-282