Learn More
Medchemexpress LLC DL-Proline-d7 | 65807-21-8 | 98.0% | 122.17 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW012908S100MG
Encompass_Preferred
DL-Proline-d7 is the deuterium-labeled form of DL-Proline (HY-W012908). It serves as a biochemical reagent suitable for use as a biological material or organic compound in life science-related research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes into drug molecules aids in quantitation during the drug development process.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54078