Learn More
Medchemexpress LLC DL-Pyroglutamic acid-d5 | 352431-30-2 | 99.2% | 134.14 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW012738S10MG
Encompass_Preferred
DL-Pyroglutamic acid-d5 is a deuterium-labeled form of DL-Pyroglutamic acid. This compound is valuable in research as a tracer and internal standard for quantitative analysis. Deuteration, the process of incorporating stable heavy isotopes like deuterium, can influence the pharmacokinetic and metabolic profiles of drugs, making this compound useful in drug development studies.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable for NMR, GC-MS, or LC-MS analysis
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-55043