Learn More
Medchemexpress LLC HY-107522 5mg Medchemexpress, DL-TBOA CAS:205309-81-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1075225MG
Encompass_Preferred
Medchemexpress, HY-107522 5mg DL-TBOA CAS:205309-81-5 DL-TBOA is a potent non-transportable inhibitor of excitatory amino acid transporters with IC50s of 70 μM, 6 μM and 6 μM for excitatory amino acid transporter-1 (EAAT1), EAAT2 and EAAT3, respectively. DL-TBOA inhibits the uptake of [14C]glutamate in COS-1 cells expressing the human EAAT1 and EAAT2 with Ki valuesof 42 μM and 5.7 μM, respectively. DL-TBOA blocks EAAT4 and EAAT5 in a competitive manner with Ki values of 4.4 μM and 3.2 μM, respectively[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8429