Learn More
Medchemexpress LLC DL-threo-2-methylisocitrate sodium | 71183-66-9 | 95.0% | 544.19 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY16581A1MG
Encompass_Preferred
DL-threo-2-methylisocitrate sodium salt is a substrate of isocitrate lyase 1 (ICL1).
  • For research use only.
  • Not sold to patients.
  • Solid, white to off-white appearance.
  • Soluble in H2O: 100 mg/mL (183.76 mM; requires ultrasonication).
Catalog No. 50-003-93872