Learn More
Cayman Chemical DL-threo-IsocItrIc AcIdodI 1g
SDP
Supplier:  Cayman Chemical 303781 G
Encompass_Preferred
A racemic mixture of D-threo-isocitric acid and L-threo-isocitric acid; has been used as a substrate to measure the activity of yeast isocitrate lyase in C. albicans, S. cerevisiae, or K. lactis cell-free extracts
Catalog No. 50-299-2471