Learn More
Medchemexpress LLC DL-threo-PDMP hydroc 10mM 1mL | 80938-69-8 | 427.02 | C23H39ClN2O3 | 1 ML

Supplier: Medchemexpress LLC HY116392B10MM 1ML
DL-threo-PDMP hydrochloride is the hydrochloride salt of DL-threo-PDMP, a competitive inhibitor of glucosylceramide synthase used to block glycosphingolipid biosynthesis in research. It is supplied as a 10 mM solution in DMSO (1 mL) for use in cellular and biochemical assays.
- Competitive inhibitor of glucosylceramide synthase (GCS).
- Supplied as a 10 mM solution in DMSO, 1 mL.
- High reported purity of approximately 99.4%.
- Molecular weight 427.02 g/mol and formula C23H39ClN2O3.
- Used for research in sphingolipid metabolism and related cellular assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.