Learn More
Medchemexpress LLC DL-Tryptophan | 54-12-6 | 99.98% | 204.23 | 1 G

Supplier: Medchemexpress LLC HYW0124801G
DL-Tryptophan is an endogenous metabolite for research use only. It is a white to light yellow solid.
- Endogenous metabolite
- Soluble in DMSO
- Not for medical applications
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.