Learn More
Medchemexpress LLC DL-Tryptophan | 54-12-6 | 99.98% | 204.23 | 10 G

Supplier: Medchemexpress LLC HYW01248010G
DL-Tryptophan is an endogenous metabolite.
- Appearance: Solid
- Color: White to light yellow
- Shipping: Room temperature in continental US
- Storage (powder): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.