Learn More
Medchemexpress LLC DL-Tryptophan-d3 | 340257-61-6 | 99.3% | 207.24 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW012480S5MG
Encompass_Preferred
DL-Tryptophan-d3 is the deuterium labeled DL-Tryptophan, an endogenous metabolite. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-53593