Learn More
Medchemexpress LLC HY-Y0123 500mg Medchemexpress, DL-Tyrosine CAS:556-03-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0123500MG
Encompass_Preferred
Medchemexpress, HY-Y0123 500mg DL-Tyrosine CAS:556-03-6 DL-Tyrosine is an aromatic nonessential amino acid synthesized from the essential amino acid phenylalanine. DL-Tyrosine is a precursor for several important neurotransmitters (epinephrine, norepinephrine, dopamine). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3522