Learn More
Eurofins Discoverx PathHunter DLD1 HT6 β-Arrestin Cell Line, 2 vials. (93-0294C13)
Supplier:  Eurofins Discoverx 930294C13
Encompass
The PathHunter HTR6 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin/5-HT) based activation of HTR6 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR6 and EA-tagged β-Arrestin-2. Activation of HTR6 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0190
Add to Cart
Request a Quote