Learn More
Eurofins Discoverx PathHunter DLD1 mPTGER4 β-Arrestin Cell Line, 2 vials. (93-0982C13)
Supplier:  Eurofins Discoverx 930982C13
Encompass
The PathHunter mPTGER4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin E2) based activation of mPTGER4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mPTGER4 and EA-tagged β-Arrestin-2. Activation of mPTGER4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1741
Add to Cart
Request a Quote