Learn More
Cayman Chemical DLIn-MC4-DMA 50mg
SDP
Supplier:  Cayman Chemical 3973250 MG
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.93); LNPs containing DLin-MC4-DMA and encapsulating siRNA for F7 reduce its gene expression levels in mice
Catalog No. 50-294-2603