Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dlin-MeOH | 1169768-28-8 | 98.0% | C37H68O | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY142986

Encompass_Preferred

Dlin-MeOH is a lipid product developed for use in drug delivery systems. It is intended for research purposes only and is not sold to patients.

  • Purity of 98.0%
  • Molecular weight of 528.94
  • Appears as a colorless to light yellow liquid
  • Recommended storage for pure form: -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in solvent form: -80°C for 6 months or -20°C for 1 month
  • Suitable for drug delivery applications

Catalog No. 50-003-56721


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.