Learn More
Medchemexpress LLC DLK-1 Human HEK293 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7791450UG
Encompass_Preferred
The DLK-1 protein has been implicated in neuroendocrine differentiation suggesting a role in the complex processes that control neuroendocrine cell development and specialization As a monomer the single molecular form of DLK-1 helps carry out its biological activity DLK-1 Protein Human (HEK293 His) is the recombinant human-derived DLK-1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87714