Learn More
ABclonal Technology DLL1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14277500UL
Encompass_Preferred
DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may play a role in cell-to-cell communication.
Catalog No. 50-001-06444