Learn More
Genscript Corporation DLL3 His Cynomolgus
Supplier:  Genscript Corporation Z03947500
Encompass
Delta-like protein 3 (DLL3) is a transmembrane protein that belongs to the Delta/Serrate/Lag-2 (DSL) family of Notch ligands DLL3 inhibits primary neurogenesis May be required to divert neurons along a specific differentiation pathway Plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm (By similarity)
Catalog No. 50-000-59454