Learn More
ABclonal Technology DLST Rabbit mAb
SDP
Supplier:  ABclonal Technology A2773820UL
Encompass_Preferred
This gene encodes a mitochondrial protein that belongs to the 2-oxoacid dehydrogenase family This protein is one of the three components (the E2 component) of the 2-oxoglutarate dehydrogenase complex that catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO(2) Alternatively spliced transcript variants have been found for this gene
Catalog No. 50-000-14131