Learn More
Cospheric LLC Density Marker Beads, Fluorescent Violet, 1.06g/cc, 20% concentration, 2.5 mL
SDP
Supplier:  Cospheric LLC DMBFVIO1.06 2.5ML
Encompass
Density marker beads are small colored microspheres of known mass density that are used for calibrating density gradients and determining density in gradient columns.
Catalog No. NC1927113