Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DM-Nitrophen | 117367-86-9 | 98.40% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY12678910MG

Encompass_Preferred

DM-Nitrophen is a Ca2+ cage that releases Ca2+ when cleaved upon illumination with near-ultraviolet light. It appears as a solid with a light yellow to yellow color. This chemical has a molecular weight of 473.39 and a chemical formula of C18H23N3O12.

  • Molecular weight: 473.39
  • Chemical formula: C18H23N3O12
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping conditions: Room temperature in continental US
  • Storage for powder: -20°C for 3 years
  • Storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Solubility in H2O: 25 mg/mL (52.81 mM)
  • Solubility in DMSO: 1 mg/mL (2.11 mM)

Catalog No. 50-003-42257


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.