Learn More
Medchemexpress LLC DM-nitrophen tetrasodium | 291517-40-3 | 90.0% | 561.32 g/mol | C18H19N3Na4O12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY126789A5MG
Encompass_Preferred
DM-nitrophen tetrasodium is a photocleavable calcium chelator that releases Ca2+ upon near-ultraviolet illumination, enabling precise temporal control of intracellular calcium levels for signaling studies. The tetrasodium salt improves aqueous solubility for use in biological buffers and is compatible with microinjection and permeabilized cell preparations.
  • Photoreleases Ca2+ upon near-ultraviolet illumination.
  • Enables rapid, localized increases in free calcium for signaling studies.
  • Water-soluble tetrasodium salt compatible with aqueous buffers.
  • Suitable for microinjection and use with permeabilized cells.
  • Molecular weight 561.32 g/mol; formula C18H19N3Na4O12.
  • Supplied in small research quantities for laboratory experiments.
Catalog No. 50-002-64452