Learn More
Medchemexpress LLC Dm1-peg4-dbco | >98.0% | 1198.79 | C63H80ClN5O16 | 5 MG

Supplier: Medchemexpress LLC HY1362615MG
DM1-PEG4-DBCO is a drug-linker conjugate that links the cytotoxic maytansinoid DM1 to a DBCO-PEG4-Ahx linker for bioorthogonal strain-promoted alkyne-azide cycloaddition (SPAAC). It is supplied as an off-white to light yellow solid for use in antibody-drug conjugate construction and click-chemistry conjugations.
- Click chemistry reagent with DBCO for SPAAC conjugation to azide-containing molecules.
- Contains DM1, a potent microtubule inhibitor suitable for ADC payloads.
- Soluble in DMSO at 100 mg/mL; requires ultrasonic assistance and fresh DMSO.
- Storage: store at -20°C; in solvent store at -80°C for up to 6 months or -20°C for up to 1 month; protect from light.
- Appearance: off-white to light yellow solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.