Learn More
Medchemexpress LLC HY-139441 5mg , DM21 CAS:2243689-80-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1394415MG
Encompass_Preferred
HY-139441 5mg DM21 CAS: 2243689-80-5 DM21 is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1446