Learn More
Medchemexpress LLC DM4-d6 | 796073-69-3 | 99.1% | 786.40 | 100 UG
SDP
Supplier:  Medchemexpress LLC HY12454S100UG
Encompass_Preferred
DM4-d6 is deuterium labeled DM4. DM4 is an antitubulin agent that inhibits cell division and can be used in the preparation of an antibody agent conjugate. It can also be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled DM4
  • Antitubulin agent
  • Inhibits cell division
  • Can be used in the preparation of antibody agent conjugate
  • For research use only
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-37269