Learn More
Anton Paar DMA 35 Ex Petrol Intrinsically safe portable density meter for petroleum
Supplier:  Anton Paar 183057
Encompass_Preferred
DMA 35 Ex Petrol is an intrinsically safe portable density and concentration meter (Ex marking [Ex] II 2 G Ex ib IIB T4) for use in hazardous areas designed for the petroleum industry. Its special housing is resistant to petroleum products. DMA 35 Ex Petrol fully complies with the IP 559 and ASTM D7777 standards. Only 2 mL of sample are required for a measurement, performed based on the oscillating U-tube technology. If the measuring cell nevertheless suffers from cell rupture, you benefit from the unique patented connection of the measuring cell, which allows replacement. An RFID interface for quick sample identification and a Bluetooth interface for convenient data handling in the field are integral parts of the instrument.Manual calculations or corrections via a table are things of the past- the measuring results are directly converted into API numbers. DMA 35 Ex Petrol has protection class IP54 and high resistance for use in harsh environments.Includes Carrying Case.
Catalog No. NC1818214