Learn More
Medchemexpress LLC Dmani | 2978635-30-0 | 99.8% | 482.40 g/mol | C25H27IN2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD247910MG
Encompass_Preferred
DMANI is an intramolecular charge-transfer (ICT)-based, mitochondria-targeted ratiometric fluorescent probe for selective detection of mitochondrial peroxynitrite (ONOO-) in living cells. It shows distinct absorption and emission spectral changes upon reaction with ONOO-, enabling ratiometric fluorescence imaging for live-cell studies.
  • Mitochondria-targeted ratiometric fluorescent probe
  • Detects mitochondrial peroxynitrite (ONOO-) in living cells
  • Distinct dual-wavelength spectral response for ratiometric measurement
  • High chemical purity (99.8%)
  • Solid form with recommended storage at 4°C, protected from moisture and light
  • Available in small research pack sizes, e.g. 10 mg
Catalog No. 50-002-97690