Learn More
Medchemexpress LLC DMAPT (Dimethylamino parthenolide) | 791595-09-0 | 98.0% | 293.40 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16172
Encompass_Preferred
DMAPT (Dimethylamino Parthenolide), an analogue of Parthenolide (PTL), is an oral active NF-κB inhibitor with potential anti-cancer and anti-metastatic effects. It has an LD50 of 1.7 μM for cell population in AML cells.
  • Oral active NF-κB inhibitor.
  • LD50 of 1.7 μM for cell population in AML cells.
  • Potential anti-cancer and anti-metastatic effect.
  • Decreased constitutive NF-κB binding activity and inhibits cell proliferation and viability of PC-3 and DU145 cells.
  • Increases sensitivity of PC-3 tumor xenografts to X-rays in vivo.
  • Slows normal tumor development in TRAMP mice.
  • Reduces metastatic area in lung tissues of TRAMP mice.
Catalog No. 50-003-91406