Learn More
TARGETMOL CHEMICALS INC DMAT 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T739025MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DMAT (Casein kinase II Inhibitor) is a potent and specific inhibitor of CK2(IC50 value of 130 nM). purity: 99%
Catalog No. 50-257-9199