Learn More
Apexbio Technology LLC DMAT 749234-11-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A33685.1
Encompass_Preferred
DMAT (CAS 749234-11-5) known as 2-dimethylamino-4 5 6 7-tetrabromo-benzimidazole is a small-molecule inhibitor primarily targeting protein kinase CK2 with an IC50 of 0 13 M It also inhibits other kinases notably PKD1 PIM3 and PIM1 CK2 is implicated in the regulation of cell proliferation and survival and is frequently overexpressed in various human cancers In cellular models DMAT induces caspase-mediated apoptosis in breast cancer cells resistant to antiestrogens and suppresses steroid hormone secretion and proliferation in adrenocortical carcinoma cells In vivo studies indicate that DMAT interferes with NF B and Wnt signaling pathways and impairs hepatoma cell growth by targeting CK2 and PIM-3 supporting its utility in cancer research
Catalog No. 50-000-96872