Learn More
Selleck Chemical LLC DMB E4779-1G
SDP
Supplier:  Selleck Chemical LLC E47791G
Encompass_Preferred
DMB (GLP-1R agonist 2 Compound 2) is an agonist of the GLP-1 receptor (GLP-1R) with a Ki of 13 25 M It activates GLP-1R by forming hydrogen bonds with the Tyr42 Cys71 and Ser84 residues DMB attenuates transient focal cerebral ischemia injury and inhibits neuronal apoptosis induced by MCAO
Catalog No. 50-000-39585