Learn More
Apexbio Technology LLC DMBA-250mg
SDP
Supplier:  Apexbio Technology LLC C8730250
Encompass
DMBA (CAS No 57-97-6) is a chemical compound commonly studied in the field of biomedical research notably for its role as a polycyclic aromatic hydrocarbon Originating from both natural sources and anthropogenic activities it is crucial in exploring carcinogenic processes DMBA functions by inducing DNA adduct formation leading to mutagenesis which makes it a valuable tool in understanding cancer initiation and progression In vitro DMBA exhibits activity typically in the nanomolar to low micromolar range demonstrating its potency It is frequently used in various experimental models including cell lines and animal studies to investigate carcinogenesis and the efficacy of potential chemopreventive agents Additionally its role in drug screening for anti-cancer properties is significant Experimental concentrations typically depend on the specific research design ensuring precise and relevant findings
Catalog No. 50-005-67934