Learn More
Medchemexpress LLC Dmba-sil-mal-mmae | 2923885-49-6 | 1403.65 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16070250MG
Encompass_Preferred
DMBA-SIL-Mal-MMAE is a cytotoxin-linker conjugate for ADC with potent antitumor activity, utilizing the anti-mitotic agent monomethyl auristatin E (MMAE), a tubulin inhibitor, linked DMBA-SIL-Mal. This product is for research use only and not sold to patients.
  • Cytotoxin-linker conjugate for ADC
  • Potent antitumor activity
  • Uses monomethyl auristatin E (MMAE)
  • MMAE is a tubulin inhibitor
Catalog No. 50-003-95194