Learn More
Medchemexpress LLC Dmba-sil-mal-mmae | 2923885-49-6 | >98.0% | 1403.65 | C72H106N8O20 | 10 MG

Supplier: Medchemexpress LLC HY16070210MG
A cytotoxin-linker conjugate pairing the antimitotic agent monomethyl auristatin E (MMAE) with a DMBA-SIL-Mal linker for use in antibody-drug conjugate (ADC) research, synthesis, and analytical characterization.
- Contains monomethyl auristatin E (MMAE) tubulin inhibitor payload.
- Designed for ADC synthesis and analytical characterization.
- Available in multiple research-scale sizes, including 10 mg.
- Recommended storage: sealed at -20°C; in solvent, -80°C for long-term storage.
- Molecular weight 1403.65 and molecular formula C72H106N8O20.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.