Learn More
TARGETMOL CHEMICALS INC DMCM HYDROCHLORIDE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1106110MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DMCM hydrochloride is a non-selective fully inverse agonist of benzodiazepine. DMCM showed significant differences in human recombinant GABAA (alpha)x(beta)3(gamma)2 receptor subtypes. For (alpha)1 (alpha)2 (alpha)3 and (alpha)5 receptors their Kis were 10 nM 13 nM 7.5 nM 2.2 nM respectively. purity: 98%
Catalog No. 50-258-1583