Learn More
Medchemexpress LLC DMCM hydrochloride | 1215833-62-7 | 99.1% | 350.80 g·mol⁻¹ | C17H19ClN2O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100369A10MM 1ML
Encompass_Preferred
DMCM hydrochloride is a benzodiazepine-site inverse agonist supplied as a ready-to-use 10 mM solution in DMSO for research applications. It is commonly used in pharmacology and neuroscience to probe GABAA receptor function and subtype pharmacology, offering consistent concentration and high chemical purity for in vitro assays. Intended for laboratory research use only.
  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity (≈99.1%) suitable for biochemical assays.
  • Well-characterized GABAA receptor inverse agonist activity for subtype profiling.
  • Stable solution format reduces preparation time and variability.
  • Supplied with CAS and molecular identifier data for unambiguous identification.
Catalog No. 50-002-14287