Learn More
Cell Biologics DMEM 500MLX6
SDP
Supplier:  Cell Biologics M6216
Encompass_Preferred
DMEM,with High Glucose, L-glutamine, Sodium Pyruvate and, 25mM HEPES Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME), containing four-fold concentrations of the amino acids and vitamins found in BME, as well as additional supplementary components. The original formulation of DMEM contained 1000 mg/L of glucose, but has since been raised in some modifications to 4500 mg/L, giving the terms "Low Glucose" and "High Glucose". Several other modifications over the years, in addition to the alternative glucose levels, have broadened the spectrum of cell types supported by DMEM
Catalog No. NC1917076