Learn More
ATCC DMEM: F-12 Medium
Supplier:  ATCC 302006
Encompass
DMEM: F-12 Medium contains 2.5 mM L-glutamine, 15 mM HEPES, 0.5 mM sodium pyruvate, and 1200 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-3812
 Restricted Item